Synonym:
2,2′-(1,8-Dihydroxy-3,6-disulfonaphthylene-2,7-bisazo)bisbenzenearsonic acid,
2,7-
CAS Number 1668-00-4
Linear Formula (HO)2C10H2(SO3H)2(N=NC6H4AsO3H2)2
Molecular Weight 776.37
Beilstein Registry Number 5717957
EC Number 216-788-6
MDL number MFCD00036695
PubChem Substance ID 24846864
| Related Categories | Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More... |
| quality | for spectrophotometric det. of Th, Zr, U, Cd, Zn, Ca |
| mp | >320 °C(lit.) |
| cation traces | Ca: ≤500 mg/kg |
| Cd: ≤50 mg/kg | |
| Cr: ≤200 mg/kg | |
| Mg: ≤50 mg/kg | |
| Mo: ≤50 mg/kg |
Visit our Titration Center to learn more.
Reagent for the determination of Th, U, Zr and rare earth metals1,2; Calcium indicator dye3
Customers Also Viewed
low viscosity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?