EMAIL THIS PAGE TO A FRIEND

11105 Fluka

Artemisia ketone

purum, ≥97.0% (GC)

Synonym: 3,3,6-Trimethyl-1,5-heptadien-4-one

  • CAS Number 546-49-6

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 1701720

  •  EC Number 208-903-3

  •  MDL number MFCD00151948

  •  PubChem Substance ID 24847006

Purchase

Properties

grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.466
density   0.869 g/mL at 20 °C(lit.)

Price and Availability

Customers Also Viewed

(+)-Aromadendrene

≥97.0% (sum of enantiomers, GC)

Bornyl acetate

≥98%, FG

Sodium methanethiolate

technical, ≥90% (RT)

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1224C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 1,IV,3571

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?