Synonym: Nonanedioic acid
CAS Number 123-99-9
Linear Formula HO2C(CH2)7CO2H
Molecular Weight 188.22
Beilstein Registry Number 1101094
EC Number 204-669-1
MDL number MFCD00004432
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis, |
| vapor density | 6.5 (vs air) |
| vapor pressure | <1 mmHg ( 20 °C) |
| grade | technical |
| assay | ~85% (GC) |
| bp | 286 °C/100 mmHg(lit.) |
| mp | 109-111 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?