Synonym: Benzyldimethyldodecylammonium bromide
CAS Number 7281-04-1
Linear Formula CH3(CH2)11N(Br)(CH3)2CH2C6H5
Molecular Weight 384.44
Beilstein Registry Number 3579193
EC Number 230-698-4
MDL number MFCD00011768
Popular Documents: FTNMR (PDF)
| Related Categories | Ammonium Salts, Cationic Detergents, Chemical Synthesis, Detergents, Detergents A to M, |
| description | cationic |
| grade | purum |
| assay | ≥97.0% (AT) |
| mp | 50-55 °C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?