Synonym: Benzyltetradecyldimethylammonium chloride
CAS Number 139-08-2
Linear Formula CH3(CH2)13N(Cl)(CH3)2CH2C6H5
Molecular Weight 368.04
Beilstein Registry Number 4062599
EC Number 205-352-0
MDL number MFCD00011771
PubChem Substance ID 24848127
| Related Categories | Ammonium, Ammonium Salts, Cationic Detergents, Chemical Synthesis, Detergents, |
| description | cationic |
| assay | ≥99.0% (AT) |
| quality | anhydrous |
| mp | 56-62 °C |
10, 50 g in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?