CAS Number 4568-71-2
Empirical Formula (Hill Notation) C13H16ClNOS
Molecular Weight 269.79
Beilstein Registry Number 3788921
EC Number 224-947-6
MDL number MFCD00011959
PubChem Substance ID 24848161
Popular Documents: FTIR (PDF)
| Related Categories | Chemical Synthesis, Heterocyclic Ammonium Salts, Phase Transfer Catalysts, Synthetic Reagents |
| grade | purum |
| assay | ≥99.0% (AT) |
| mp | 142-144 °C(lit.) |
Catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters1,2,3
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?