Synonym: ‘Triglycol dichloride’, Dichlorotriethylene dioxide, Tri(ethylene glycol) dichloride
CAS Number 112-26-5
Linear Formula ClCH2CH2OCH2CH2OCH2CH2Cl
Molecular Weight 187.06
Beilstein Registry Number 506177
EC Number 203-952-7
MDL number MFCD00000976
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, C2 to C7, Chemical Synthesis, Ethers, Organic Building Blocks, |
| grade | purum |
| assay | ≥95.0% (GC) |
| impurities | ~0.5% water |
| refractive index |
n |
|
n | |
| bp | 235 °C(lit.) |
| density | 1.197 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?