• USA Home
  • 14715 - (R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine-manganese(III) chloride

EMAIL THIS PAGE TO A FRIEND

14715 Fluka

(R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine-manganese(III) chloride

purum, ≥96.0% (CHN)

Synonym: (R,R)-Jacobsen’s catalyst

Purchase

Properties

grade   purum
assay   ≥96.0% (CHN)

Description

Other Notes

Catalyst for the enantioselective epoxidation of olefins lacking functional groups1,2; Catalyst for enantioselective hydrolysis of epoxides3

Price and Availability

Safety & Documentation

Safety Information

Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. S. Chang et al. Tetrahedron 58, 6939, (1993)

2. E.N. Jacobsen et al. Tetrahedron 50, 4323, (1994)

3. Practical Access to Highly Enantioenriched C-3 Building Blocks via Hydrolytic Kinetic Resolution. M.E. Furrow et al. J. Org. Chem. 63, 6776, (1998)

Loading...


Merck 14,5252

Related Products

replaced by

Product #

Description

Add to Cart

404446 (R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?