Synonym:
N-Boc-4-piperidone,
tert-Butyl 4-
CAS Number 79099-07-3
Empirical Formula (Hill Notation) C10H17NO3
Molecular Weight 199.25
Beilstein Registry Number 3650236
MDL number MFCD00151800
PubChem Substance ID 24849492
| Related Categories | Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Piperidones |
| grade | puriss. |
| assay | ≥99.0% (HPLC) |
| mp | 73-77 °C(lit.) |
| 73-77 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?