Synonym:
(+)-Bornanamine,
endo-(1R)-
CAS Number 32511-34-5
Empirical Formula (Hill Notation) C10H19N
Molecular Weight 153.26
Beilstein Registry Number 2801831
EC Number 251-076-9
MDL number MFCD00042691
PubChem Substance ID 24849605
Popular Documents: FTNMR (PDF)
| Related Categories | Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More... |
| grade | purum |
| assay | ≥97.0% (NT) |
| optical activity | [α]20/D +46±2°, c = 0.5% in ethanol |
| mp | 160-163 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?