EMAIL THIS PAGE TO A FRIEND

16000 Fluka

Pyruvonitrile

technical, ≥90% (GC)

Synonym: 2-Oxopropionitrile, Acetyl cyanide

  • CAS Number 631-57-2

  • Linear Formula CH3COCN

  • Molecular Weight 69.06

  •  Beilstein Registry Number 1737633

  •  MDL number MFCD00001838

  •  PubChem Substance ID 24849842

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.376(lit.)
  n20/D 1.376
bp   92-93 °C(lit.)
density   0.974 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability


Search for Answers, Not Just Materials

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
16-45-61
RIDADR 
UN 3273 6.1(3) / PGII
WGK Germany 
3
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 3,IV,1515

Corp MSDS 1 (2), 3018:B / FT-IR 2 (1), 1456:B / FT-IR 1 (1), 862:A / FT-NMR 1 (1), 1385:B / RegBook 1 (1), 1009:F / Structure Index 1, 157:C:6 / Vapor Phase 3, 816:C

Related Products

replaced by

Product #

Description

Add to Cart

269212 Pyruvonitrile, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?