EMAIL THIS PAGE TO A FRIEND

16300 Fluka

4-Bromobenzaldehyde

purum, ≥97.0% (GC)

Purchase

Properties

grade   purum
assay   ≥97.0% (GC)
impurities   ≤2% 4-bromobenzoic acid
mp   55-57 °C
  55-58 °C(lit.)

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37
WGK Germany 
2
RTECS 
CU4810000
Flash Point(F) 
228 °F
Flash Point(C) 
109 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 7,IV,579

Aldrich MSDS 1, 231:A / Corp MSDS 1 (1), 500:B / FT-IR 1 (2), 110:D / FT-IR 2 (2), 2529:C / FT-NMR 1 (2), 940:C / IR-Spectra (3), 914:F / IR-Spectra (2), 800:E / NMR-Reference 2 (2), 109:B / RegBook 1 (2), 1717:G / Sigma FT-IR 1 (2), 332:C / Structure Index 1, 272:D:6 / Vapor Phase 3, 1290:B

Related Products

replaced by

Product #

Description

Add to Cart

B57400 4-Bromobenzaldehyde, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?