• USA Home
  • 16355 - Nitric acid concentrate

EMAIL THIS PAGE TO A FRIEND

16355 Fluka

Nitric acid concentrate

0.1 M HNO3 in water (0.1N), eluent concentrate for IC

Synonym: Nitric acid solution

  • CAS Number 7697-37-2

  • Empirical Formula (Hill Notation) HNO3

  • Molecular Weight 63.01

  •  MDL number MFCD00011349

  •  PubChem Substance ID 24849754

  •  eCl@ss 38060304

Purchase

Description

General description

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.
Download at: http://www.sigma-aldrich.com.

Preparation Note

Prepared with nitric acid and high purity water (18.2 MΩ, 0.2 μm filtered)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

2,6-<WBR>Pyridinedicarboxylic acid concentrate

0.02 M C7H5NO4 in water (0.04N), eluent concentrate for IC

Nitric acid concentrate

for 1L standard solution, 0.1 M HNO3 (0.1N)

Nitric acid concentrate

for 1L standard solution, 1.0 M HNO3 (1.0N)

Nitric acid solution

volumetric, 0.1 M HNO3 (0.1N)

Nitric acid solution

volumetric, 1 M HNO3 (1N)

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Photochemical renoxification of nitric acid on real urban grime. Baergen AM and Donaldson DJ Environ. Sci. Technol. 47(2), 815-20, (2013)

Loading...


Hair as a biomarker of environmental manganese exposure. Eastman RR, Jursa TP, Benedetti C, et al. Environ. Sci. Technol. 47(3), 1629-37, (2013)

Loading...


Lead recovery from cathode ray tube funnel glass with mechanical activation. Yuan W, Li J, Zhang Q, et al. J. Air Waste Manag. Assoc. 63(1), 2-10, (2013)

Loading...


Determination of cadmium in biodiesel using microemulsion and electrothermal atomization atomic absorption spectrometry. Lima AS, Silva DG, and Teixeira LS Environ. Monit. Assess. 187(1), 4122, (2015)

Loading...


Determination of vanadium in groundwater samples with an improved kinetic spectrophotometric method. Bağda E Environ. Technol. 35(9-12), 1165-74, (2014)

Loading...


Reducing NO(x) emissions from a nitric acid plant of domestic petrochemical complex: enhanced conversion in conventional radial-flow reactor of selective catalytic reduction process. Abbasfard H, Hashemi SH, Rahimpour MR, et al. Environ. Technol. 34(17-20), 2867-79, (2013)

Loading...


[Case of dyspnea due to toilet cleaner containing nitric acid for household use]. Yanagawa Y, Matsukawa T, Yokoyama K, et al. Chudoku. Kenkyu. 26(3), 240-3, (2013)

Loading...


Development of a method for recycling of CRT funnel glass. Ling TC and Poon CS Environ. Technol. 33(22-24), 2531-7, (2012)

Loading...


A novel approach for preferential recovery of Sr from (Sr, Th)O2. Vyas CK, Joshirao PM, Shukla R, et al. J. Hazard. Mater. 241-242, 340-7, (2012)

Loading...


Effect of surface modification on single-walled carbon nanotube retention and transport in saturated and unsaturated porous media. Tian Y, Gao B, Morales VL, et al. J. Hazard. Mater. 239-240, 333-9, (2012)

Loading...


Photochemistry of molecules relevant to the atmosphere: photodissociation of nitric acid in the gas phase. Huber JR ChemPhysChem 5(11), 1663-9, (2004)

Loading...


Optimization of selenylation conditions for Chinese angelica polysaccharide based on immune-enhancing activity. Qin T, Chen J, Wang D, et al. Carbohydr. Polym. 92(1), 645-50, (2013)

Loading...


A comparative study of heavy metal accumulation and antioxidant responses in Vaccinium myrtillus L. leaves in polluted and non-polluted areas. Kandziora-Ciupa M, Ciepał R, Nadgórska-Socha A, et al. Environ. Sci. Pollut. Res. Int. 20(7), 4920-32, (2013)

Loading...


Application of the extraction induced by emulsion breaking for the determination of chromium and manganese in edible oils by electrothermal atomic absorption spectrometry. Robaina NF, Brum DM, and Cassella RJ Talanta 99, 104-12, (2012)

Loading...


Speciation in aqueous solutions of nitric acid. Hlushak S, Simonin JP, De Sio S, et al. Dalton Trans. 42(8), 2853-60, (2013)

Loading...


Poly(methacrylic acid)-grafted chitosan microspheres via surface-initiated ATRP for enhanced removal of Cd(II) ions from aqueous solution. Huang L, Yuan S, Lv L, et al. J. Colloid. Interface Sci. 405, 171-82, (2013)

Loading...


Microwave-assisted digestion of organic samples: how simple can it become? Nóbrega JA, Pirola C, Fialho LL, et al. Talanta 98, 272-6, (2012)

Loading...


Atmospheric acid gases in Tokushima, Japan, monitored with parallel plate wet denuder coupled ion chromatograph. Takeuchi M, Miyazaki Y, Tsunoda H, et al. Anal. Sci. 29(1), 165-8, (2013)

Loading...


Distinct types of tumors exhibit differential grade of inflammation and angiogenesis in mice. Viana CT, Campos PP, Carvalho LA, et al. Microvasc. Res. 86, 44-51, (2013)

Loading...


Spectroscopic study of HNO3 dissociation on ice. Marchand P, Marcotte G, and Ayotte P J. Phys. Chem. A 116(49), 12112-22, (2012)

Loading...


Cloud point extraction and flame atomic absorption spectrometry determination of lead (II) in environmental and food samples. Soylak M, Yilmaz E, Ghaedi M, et al. J. AOAC Int. 95(6), 1797-802, (2012)

Loading...


Soft cutting of single-wall carbon nanotubes by low temperature ultrasonication in a mixture of sulfuric and nitric acids. Shuba MV, Paddubskaya AG, Kuzhir PP, et al. Nanotechnology 23(49), 495714, (2012)

Loading...


Silica gel-mediated organic reactions under organic solvent-free conditions. Onitsuka S, Jin YZ, Shaikh AC, et al. Molecules 17(10), 11469-83, (2012)

Loading...


Nitrogen isotopes in ice core nitrate linked to anthropogenic atmospheric acidity change. Geng L, Alexander B, Cole-Dai J, et al. Proc. Natl. Acad. Sci. U. S. A. 111(16), 5808-12, (2014)

Loading...


Adsorption and desorption of uranium (VI) in aerated zone soil. Li X, Wu J, Liao J, et al. J. Environ. Radioact. 115, 143-50, (2013)

Loading...


Reduction of PCDDs/PCDFs in MSWI fly ash using microwave peroxide oxidation in H2SO4/HNO3 solution. Chang YM, Dai WC, Tsai KS, et al. Chemosphere 91(6), 864-8, (2013)

Loading...


The effect of ultrasonic and HNO3 treatment of activated carbon from fruit stones on capacitive and pseudocapacitive energy storage in electrochemical supercapacitors. Venhryn BY, Stotsko ZA, Grygorchak II, et al. Ultrason. Sonochem. 20(5), 1302-7, (2013)

Loading...


Proton transfer reactions between nitric acid and acetone, hydroxyacetone, acetaldehyde and benzaldehyde in the solid phase. Lasne J, Laffon C, and Parent P Phys. Chem. Chem. Phys. 14(45), 15715-21, (2012)

Loading...


Photosynthetic and antioxidant responses of Liquidambar formosana and Schima superba seedlings to sulfuric-rich and nitric-rich simulated acid rain. Chen J, Wang WH, Liu TW, et al. Plant Physiol. Biochem. 64, 41-51, (2013)

Loading...


Elemental composition of dog foods using nitric acid and simulated gastric digestions. Kelly DG, White SD, and Weir RD Food Chem. Toxicol. 55, 568-77, (2013)

Loading...


Sensitivity analyses of factors influencing CMAQ performance for fine particulate nitrate. Shimadera H, Hayami H, Chatani S, et al. J. Air Waste Manag. Assoc. 64(4), 374-87, (2014)

Loading...


Effect of surface treatment on bond strength of Ti-10Ta-10Nb to low-fusing porcelain. Lee BA, Kim OS, Vang MS, et al. J. Prosthet. Dent. 109(2), 95-105, (2013)

Loading...


Observed levels and trends of gaseous SO2 and HNO3 at Mt. Waliguan, China: results from 1997 to 2009. Lin W, Xu X, Yu X, et al. J. Environ. Sci. (China) 25(4), 726-34, (2013)

Loading...


Comparative study on corrosion behaviour of Nitinol and stainless steel orthodontic wires in simulated saliva solution in presence of fluoride ions. Mirjalili M, Momeni M, Ebrahimi N, et al. Mater. Sci. Eng. C. Mater. Biol. Appl. 33(4), 2084-93, (2013)

Loading...


Assessment of the efficacy of a new formulation for plantar wart mummification: new experimental design and human papillomavirus identification. Viennet C, Gheit T, Muret P, et al. Clin. Exp. Dermatol. 38(1), 85-8, (2013)

Loading...


Stability of aflatoxins in solution. Diaz GJ, Cepeda SM, and Martos PA J. AOAC Int. 95(4), 1084-8, (2012)

Loading...


Merck 14,6577

Fieser 1,733 / Fieser 3,212 / Fieser 4,356 / Fieser 5,474

Corp MSDS 1 (2), 2546:B / RegBook 1 (3), 3247:F / Sax 6, 2002

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?