| Related Categories | 2000/60/EC, AAS, AAS CRMs, Allergens, Alphabetical Index of Analytical Standards, |
| grade | TraceCERT® |
| concentration | 1000 mg/L Hg in nitric acid |
1000 mg/L Hg in 12% nitric acid, prepared with high purity Hg metal, HNO3 TraceSELECT® and water TraceSELECT®Ultra
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34. This CRM is traceable to SI unit kg and measured against NIST- and BAM-CRM.
Certified content incl. uncertainty and expiry date are given on label.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
TraceSELECT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
TraceCERT®, 1000 mg/L As in nitric acid
TraceCERT®, 1000 mg/L Cd in nitric acid
TraceCERT®, 1000 mg/L Cu in nitric acid
TraceCERT®, 1000 mg/L Pb in nitric acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?