• USA Home
  • 16920 - 4-Bromo-N,N-dimethylaniline

EMAIL THIS PAGE TO A FRIEND

16920 Fluka

4-Bromo-N,N-dimethylaniline

purum, ≥98.0% (T)

Purchase

Properties

grade   purum
assay   ≥98.0% (T)
bp   264 °C(lit.)
mp   52-54 °C(lit.)
  53-55 °C
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by 242950-ALDRICH | 4-Bromo-N,N-dimethylaniline 97%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
BW9300000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 12,IV,1499

Aldrich MSDS 1, 248:B / Corp MSDS 1 (1), 525:D / FT-IR 2 (2), 2050:B / FT-IR 1 (1), 1261:D / FT-NMR 1 (2), 482:B / NMR-Reference 2 (1), 1011:C / RegBook 1 (1), 1397:F / Structure Index 1, 217:D:1 / Vapor Phase 3, 1127:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?