Synonym: Neopentyl bromide
CAS Number 630-17-1
Linear Formula (CH3)3CCH2Br
Molecular Weight 151.04
Beilstein Registry Number 1730989
EC Number 211-132-5
MDL number MFCD00000209
PubChem Substance ID 24850267
Popular Documents: FTNMR (PDF)
| Related Categories | Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More... |
| grade | purum |
| assay | ≥98.0% (GC) |
| refractive index |
n |
|
n | |
| bp | 105-106 °C/767 mmHg(lit.) |
| density | 1.199 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?