-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Ammonia aqueous Ammonia water
CAS Number 1336-21-6
Linear Formula NH4OH
Molecular Weight 35.05
Beilstein Registry Number 3587154
MDL number MFCD00066650
PubChem Substance ID 24850736
| Related Categories | Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents, HPLC, |
| grade | for HPLC |
| expl. lim. | 27 % |
| concentration | ~10% NH3 in H2O (T) |
| ~10% in H2O | |
| density | 0.9 g/mL at 25 °C(lit.) |
| λ | neat |
| UV absorption | λ: 230 nm Amax: 0.1 |
Save 20% on our wide range of HPLC accessoriesCustomers Also Purchased
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?