EMAIL THIS PAGE TO A FRIEND
17843 Fluka

Ammonium formate

for HPLC, ≥99.0%

Synonym: Formic acid ammonium salt

  • CAS Number 540-69-2

  • Linear Formula HCO2NH4

  • Molecular Weight 63.06

  •  Beilstein Registry Number 3625095

  •  EC Number 208-753-9

  •  MDL number MFCD00013103

  •  PubChem Substance ID 24850743

  •  eCl@ss 39021903

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents, HPLC,
grade   for HPLC
assay   ≥99.0% (dried material, NT)
  ≥99.0%
impurities   ≤2% water
pH   5.5-7.5 (20 °C, 10% in H2O)
mp   119-121 °C(lit.)
solubility   H2O: soluble0.1 g/mL at 25 °C, clear, colorless
density   1.26 g/mL at 25 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 250 nm Amax: 0.05
  λ: 260 nm Amax: 0.04
  λ: 270 nm Amax: 0.03
  λ: 280 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds for filter test
  corresponds to standard for RP gradient test

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Application

Ammonium formate was used as buffer solution during LC-MS/MS analysis for screening of drugs and exogenous compounds. It may be used as a catalytic hydrogen transfer agent in reduction of aliphatic and aromatic nitro compounds.

General description

Ammonium formate is a ammonium salt of formic acid. It is readily available, is stable and nontoxic in nature, can be used in conjugation with Pd-C or Raney-Nickel catalysts.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
BQ6650000
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?