17850 Fluka

Ciprofloxacin

≥98.0% (HPLC)

DOWNLOAD MSDS (PDF)

Synonym: Ciprobay

  • CAS Number 85721-33-1

  • Empirical Formula (Hill Notation) C17H18FN3O3

  • Molecular Weight 331.34

  •  Beilstein Registry Number 3568352

  •  MDL number MFCD00185755

  •  PubChem Substance ID 24850750

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Price and Availability

Customers Also Viewed

Ciprofloxacin

pharmaceutical secondary standard; traceable to USP and PhEur

Ciprofloxacin-d<SUB>8</SUB> hydrochloride hydrate

VETRANAL, analytical standard

Levofloxacin

≥98.0% (HPLC)

Piperacillin sodium salt

penicillin analog

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?