17933 Fluka

Hydrogen chloride – 2-propanol solution

puriss. p.a., for GC, ~1.25 M (T)

DOWNLOAD MSDS (PDF)

Synonym: Hydrochloric acid in 2-propanol

  • Linear Formula HCl · CH3CH(OH)CH3

  • Molecular Weight 36.46

  •  Beilstein Registry Number 1098214

  •  MDL number MFCD00011324

  •  PubChem Substance ID 24850804

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Alkylation & Esterification More...
grade   for GC
  puriss. p.a.
concentration   ~1.25 M (T)
density   0.821 g/mL at 20 °C
storage temp.   2-8°C

Description

Application

Derivatizing agent for GC

Price and Availability

Customers Also Viewed

Hydrogen chloride - 1-butanol solution

~3 M in 1-butanol, for GC derivatization

Hydrogen chloride – 2-propanol solution

0.1 M HCl in 2-propanol (0.1N), for Total Base Number, according to DIN 51558-1, volumetric

Hydrogen chloride – ethanol solution

~1.25 M HCl, for GC derivatization

Hydrogen chloride – methanol solution

~1.25 M HCl, for GC derivatization

Hydrogen chloride solution

2.0 M in diethyl ether

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?