17934 Fluka

Hydrogen chloride – ethanol solution

~1.25 M HCl, for GC derivatization

DOWNLOAD MSDS (PDF)

Synonym: Hydrochloric acid in ethanol

  • CAS Number 7647-01-0

  • Linear Formula HCl · CH3CH2OH

  • Molecular Weight 36.46

  •  Beilstein Registry Number 1098214

  •  MDL number MFCD00011324

  •  PubChem Substance ID 24850806

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Alkylation & Esterification More...
grade   for GC derivatization
concentration   ~1.25 M HCl
density   0.825 g/mL at 20 °C
storage temp.   2-8°C

Description

Application

Derivatizing agent for GC

Price and Availability

Customers Also Viewed

Hydrogen chloride – 2-propanol solution

puriss. p.a., for GC, ~1.25 M (T)

Hydrogen chloride – methanol solution

~1.25 M HCl, for GC derivatization

Hydrogen chloride solution

2.0 M in diethyl ether

Hydrogen chloride solution

4.0 M in dioxane

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?