EMAIL THIS PAGE TO A FRIEND

17983 Fluka

Disperse Orange 3

for microscopy

Synonym: 4-(4-Nitrophenylazo)aniline

  • CAS Number 730-40-5

  • Linear Formula O2NC6H4N=NC6H4NH2

  • Molecular Weight 242.23

  •  Beilstein Registry Number 1842907

  •  Colour Index Number 11005

  •  EC Number 211-984-8

  •  MDL number MFCD00007856

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   for microscopy
mp   ~200 °C (dec.)(lit.)
absorption   λmax 443 nm

Description

Features and Benefits

Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength.1

Other Notes

Dye standard for the assay of allergy-inducing dyes in textiles 2,3

Price and Availability

Customers Also Viewed

Disperse Orange 13

Dye content 90 %

Disperse Orange 3

Dye content 90 %

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. Cherioux, F. et al. Chem. Phys. Lett. 319, 669, (2000)

2. K.L. Hatch Textile Res. J. 54, 664, 721, (1984)

3. Textile dye dermatitis. K.L. Hatch, H.I. Maibach J. Am. Acad. Dermatol. 32, 631, (1995)

Loading...


Multifunctional core-shell nanoparticles: discovery of previously invisible biomarkers. Tamburro D J. Am. Chem. Soc. 133(47), 19178-88, (2011)

Loading...


Beil. 16,311

Corp MSDS 1, 1476:D / FT-IR 2 (3), 4076:C / FT-IR 1 (2), 966:C / FT-NMR 1 (3), 547:C / IR-Spectra (3), 1450:F / RegBook 1 (2), 2739:D / Sigma FT-IR 1 (2), 132:A / Stains and Dyes Ref, 287 / Structure Index 1, 433:C:1

Related Products

replaced by

Product #

Description

Add to Cart

364797 Disperse Orange 3, Dye content 90 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?