CAS Number 12222-75-2
MDL number MFCD00280151
| Related Categories | 2002/61/EC, 67/548/EEC, All alphabetical sorted, Analytical Reagents, Analytical Standards, |
| grade | analytical standard, for food analysis |
Dye standard for the assay of allergy-releasing dyes in textiles1,2
Customers Also Viewed
for microscopy
1. Textile dye dermatitis. A review. K.L. Hatch, H.I. Maibach J. Am. Acad. Dermatol. 12, 1079, (1985)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?