• USA Home
  • 18122 - 1-Butyl-3-methylimidazolium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND

18122 Fluka

1-Butyl-3-methylimidazolium hexafluorophosphate

for catalysis, ≥98.5% (T)

Synonym: BMIMPF6

Purchase

Properties

Related Categories Chemical Synthesis, Imidazolium, Ionic Liquids, Specialty Synthesis
grade   for catalysis
assay   ≥98.5% (T)
impurities   ≤0.02% water
refractive index   n20/D 1.41
density   1.38 g/mL at 20 °C(lit.)
anion traces   bromide (Br-): ≤10 mg/kg
  chloride (Cl-): ≤10 mg/kg
  nitrate (NO3-): ≤10 mg/kg
  phosphate (PO43-): ≤30 mg/kg
  sulfate (SO42-): ≤10 mg/kg

Description

Packaging

5, 50 g in glass bottle

Application

Ionic Liquids for Catalysis

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Ionic Liquids for Catalysis

Several researchers have found that small impurities in ionic liquids exert a detrimental influence on the reaction outcome, particularly in catalysis. Dr. Annegret Stark of the Friedrich-Schiller Un...
Aldrich ChemFiles 2006, 6.9, 16.
Keywords: Catalysis, Chemfiles, Metathesis, Olefin metathesis

Ionic Liquids for Catalysis

Annegret Stark and Bernd Ondruschka, Friedrich-Schiller-University, Institute for Technical Chemistry and Environmental Chemistry, Lessingstr. 12, 07743 Jena, Germany; annegret.stark@uni-jena.de.
Annegret Stark and Bernd Ondruschka
Aldrich ChemFiles 2005, 5.6, 8.
Keywords: Catalysis, Environmental, Esterifications, Hydroformylations, Hydrogenations, Isomerizations, Metathesis, Separation, Solvents, Substitutions

Related Content

Ionic Liquids for Catalysis in Chemical Synthesis

Ionic liquids have been thoroughly investigated as solvents in most types of catalytic reactions.1-4 Their merit lies in the ease with which their properties can be tuned by varying either the anion,...
Keywords: Catalysis, Chemfiles, Metathesis, Solvents, Substitutions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?