• USA Home
  • 18562 - Zinc Standard for ICP
EMAIL THIS PAGE TO A FRIEND
18562 Fluka
Zinc Standard for ICP
TraceCERT®, 1000 mg/L Zn in nitric acid
Purchase
Description
Analysis Note
1000 mg/L Zn in 2% nitric acid, prepared with high purity Zn metal, HNO3 TraceSELECT® and water TraceSELECT®Ultra
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34. This CRM is traceable to SI unit kg and measured against NIST- and BAM-CRM.
Certified content incl. uncertainty and expiry date are given on label. Information about trace impurities are stated in the enclosed certificate (available as download: http://www.sigma-aldrich.com).
Other Notes
For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website
Legal Information
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
TraceSELECT is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Explore the New Analytical Standards Portal
Customers Also Viewed
Copper Standard for ICP
TraceCERT®, 1000 mg/L Cu in nitric acid
Manganese Standard for ICP
TraceCERT®, 1000 mg/L Mn in nitric acid
Nickel Standard for ICP
TraceCERT®, 1000 mg/L Ni in nitric acid
Vanadium Standard for ICP
TraceCERT®, 1000 mg/L V in nitric acid
Zinc Standard for ICP
TraceCERT®, 10'000 mg/L Zn in nitric acid
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
2
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
06929 Timestrip® 12 months, key chain
74831 Timestrip® 12 months
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?