CAS Number 125110-14-7
Empirical Formula (Hill Notation) C16H15N5O7S2 · 3H2O
Molecular Weight 507.50
MDL number MFCD03788802
PubChem Substance ID 24851186
| Related Categories | Analytical/Chromatography, Antibiotics, Antibiotics A to Z, Antibiotics A-F, Antibiotics by Application, |
| assay | ≥98.0% (HPLC) |
Visit our Sensor Applications portal to learn more.
Determination of ytterbium in mineral probes (e.g. Zeolith-catalysts, Xenotime,...)1
Customers Also Viewed
for microbiology
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?