19215 Fluka

Butyric acid

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 107-92-6

  • Linear Formula CH3CH2CH2COOH

  • Molecular Weight 88.11

  •  Beilstein Registry Number 906770

  •  EC Number 203-532-3

  •  MDL number MFCD00002814

  •  PubChem Substance ID 24851531

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories A-F, Acid, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alphabetical Index of Analytical Standards,
grade   analytical standard
vapor density   3.04 (vs air)
vapor pressure   0.43 mmHg ( 20 °C)
assay   ≥99.5% (GC)
autoignition temp.   824 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   10 %
refractive index   n20/D 1.398(lit.)
bp   162 °C(lit.)
mp   −6-−3 °C(lit.)
density   0.964 g/mL at 25 °C(lit.)
Gene Information   human ... HDAC1(3065)

Price and Availability

Customers Also Viewed

(<I>E</I>)-Benzaldehyde oxime

97% (mixture of cis and trans), (Z)-Benzaldehyde oxime ≤6%

Acetamide

≥99.0% (GC)

Butyric acid

≥99%

Butyric acid

≥99%

Butyric acid

natural, ≥99%, FCC, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?