| Related Categories | Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Food and Beverage, Petrochemistry, |
| shelf life | (limited shelf life, expiry date on the label) |
| mfr. no. | Paragon Scientific Ltd. |
| viscosity | 2618 mPa.s(25 °C) |
| density | 0.8781 g/mL at 25 °C |
Customers Also Viewed
D1000, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 1010 mPa.s (25 °C)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?