-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
CAS Number 7188-38-7
Linear Formula (CH3)3CNC
Molecular Weight 83.13
Beilstein Registry Number 1903156
EC Number 230-553-5
MDL number MFCD00000002
PubChem Substance ID 24851976
| grade | purum |
| assay | ≥98.0% (GC) |
| refractive index |
n |
|
n | |
| bp | 91 °C(lit.) |
| density | 0.735 g/mL at 25 °C(lit.) |
Reaction with organometallic reagents, a review1; Cyanation of acetals2
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?