21000 Fluka

Cesium hydroxide monohydrate

≥95.0% (T)

DOWNLOAD MSDS (PDF)
  • CAS Number 35103-79-8

  • Empirical Formula (Hill Notation) HCsO · H2O

  • Molecular Weight 167.93

  •  EC Number 244-344-1

  •  MDL number MFCD00010964

  •  PubChem Substance ID 24852606

Properties

Related Categories Cesium Salts, Chemical Synthesis, Inorganic Bases, Materials Science, Metal and Ceramic Science,
assay   ≥95.0% (T)
impurities   ~5% cesium carbonate
mp   272 °C(lit.)
density   3.68 g/mL at 25 °C(lit.)

Description

Packaging

10, 50, 250 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Cesium hydroxide monohydrate

99.95% trace metals basis

Cesium hydroxide monohydrate

99.9% trace metals basis

Cesium hydroxide solution

50 wt. % in H2O, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?