• USA Home
  • 21176 - Calcium hypophosphite

EMAIL THIS PAGE TO A FRIEND
21176 Fluka

Calcium hypophosphite

puriss. p.a., ≥99.0% (RT)

Synonym: Calcium phosphinate

Purchase

Properties

grade   puriss. p.a.
assay   ≥99.0% (RT)
loss   ≤1% loss on drying, 105 °C
pH   4.7-5.5 (25 °C, 50 mg/mL in H2O)
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤500 mg/kg
  Mn: ≤5 mg/kg
  NH4+: ≤10 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤50 mg/kg
  Zn: ≤5 mg/kg

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Other Notes

This product has been replaced by 04408-Sigma-Aldrich | Calcium hypophosphite, puriss., meets analytical specification of DAC, 98-100.5% | Ca(H2PO2)2

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3178 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?