Synonym:
(+)-β-Camphorsulfonic acid,
(1S)
CAS Number 3144-16-9
Empirical Formula (Hill Notation) C10H16O4S
Molecular Weight 232.30
Beilstein Registry Number 2809675
EC Number 221-554-1
MDL number MFCD00064157
PubChem Substance ID 24852789
Popular Documents: FTNMR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, |
| grade | purum |
| assay | ≥98.0% (T) |
| optical activity | [α]20/D +21.5±1°, c = 10% in H2O |
| mp | ~200 °C (dec.)(lit.) |
Reagent used for the resolution of bases; use as mobile phase additive for LC separations1,2; Reagent used for crystallization-induced asymmetric transformations3,4
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?