21529 Fluka

Hexanoic acid

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Acid C6, Caproic acid

  • CAS Number 142-62-1

  • Linear Formula CH3(CH2)4COOH

  • Molecular Weight 116.16

  •  Beilstein Registry Number 773837

  •  EC Number 205-550-7

  •  MDL number MFCD00004421

  •  PubChem Substance ID 24852882

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acid, All Alphabetically Sorted, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard
vapor density   4 (vs air)
vapor pressure   0.18 mmHg ( 20 °C)
assay   ≥99.0% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.416
  n20/D 1.4161(lit.)
bp   202-203 °C(lit.)
mp   −4 °C(lit.)
density   0.927 g/mL at 25 °C(lit.)

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Decanoic acid

analytical standard

Hexanoic acid

≥99.5%

Hexanoic acid

99%

Hexanoic acid

≥98%, FCC, FG

Hexanoic acid

natural, ≥98%, FCC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?