Synonym: Caproic acid methyl ester, Methyl caproate
CAS Number 106-70-7
Linear Formula CH3(CH2)4COOCH3
Molecular Weight 130.18
Beilstein Registry Number 1744683
EC Number 203-425-1
MDL number MFCD00009510
PubChem Substance ID 24852927
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chromatography, |
| grade | analytical standard |
| assay | ≥99.8% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
| density | 0.884 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?