MDL number MFCD00013129
PubChem Substance ID 24853050
| Related Categories | Microbiology, Microscopy Reagents and Kits, Staining Solutions, Staining Solutions for Acid Fast Staining, Staining Solutions for Simple Stains of Bacteria More... |
| grade | for microscopy (Bact.) |
| impurities | phenol |
| density | 0.99 g/mL at 20 °C |
Stain for the assay of acid-resistant rod-shaped bacteria1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?