EMAIL THIS PAGE TO A FRIEND

22091 Fluka

Tryptic Soy Agar

for microbiology

Synonym: CASO Agar, Soybean Casein digest Agar, TSA, Tryptone Soya Agar

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Bacillus, Bacteria (General Media),
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 15 g/L
  casein peptone (pancreatic), 15 g/L
  sodium chloride, 5 g/L
  soya peptone (papainic), 5 g/L
final pH   7.3±0.2 (25 °C)
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Bacteroides
  nonselective for Campylobacter
  nonselective for Candida
  nonselective for Cronobacter spp.
  nonselective for Escherichia coli and Coliforms
  nonselective for Neisseria
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Precultivation medium. Enumeration (E. coli) acc. to membrane-filter technique.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology, ready to use media in the bottle

for microbiology

for microbiology

Microbiologically tested.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Chronobacter spp. Classic and New Detection Methods

Cronobacter is a rod-shaped, motile and facultatively-anaerobic bacteria of the family Enterobacteriaceae. Originally Cronobacter spp. were listed as yellow-pigmented Enterobacter cloacae (see Table ...
Jvo Siegrist
Microbiology Focus Edition 1.2
Keywords: Absorption, Anaerobic, Diagnostic, Environmental, Enzyme-linked immunosorbent assay, Food Safety, Genetic, Help, Methods, Microbiology, Nutrition, Polymerase chain reaction, Proteomics, Type, Vitamins

On the trail of Campylobacter

The Campylobacter is one of the leading causes of human gastroenteritis. Common Campylobacter species C. jejuni, C. coli, and C. lari are responsible for most cases of campylobacteriosis1,2. However,...
Jvo Siegrist
AnalytiX Volume 8 Issue 1
Keywords: Antibiotics, Cancer, Citric Acid Cycle, Gram stain, Metabolism, Microbiology, Scanning electron microscopy

Streptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques

There are many natural sources of streptococci, including humans and diverse animals where they often colonize the mucosal surfaces of the mouth, intestinal tract, nasal passages and pharynx. The pre...
By Jvo Siegrist, Product Manager Microbiology, ivo.siegrist@sial.com
AnalytiX Volume 7 Article 3
Keywords: Diagnostic, Phosphorylations

Vibrio Detection

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater. Vibrios are facultative aerobe a...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Dehydration reaction, Diagnostic, Fermentation, Indicators, Metabolism, Microbiology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?