EMAIL THIS PAGE TO A FRIEND

22092 Fluka

Tryptic Soy Broth

for microbiology

Synonym: CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Bacillus, Bacteria (General Media),
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   casein peptone (pancreatic), 17 g/L
  dipotassium hydrogen phosphate, 2.5 g/L
  glucose, 2.5 g/L
  sodium chloride, 5 g/L
  soya peptone (papain digest.), 3 g/L
final pH   7.3±0.2 (25 °C)
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Bacteroides
  nonselective for Campylobacter
  nonselective for Candida
  nonselective for Escherichia coli and Coliforms
  nonselective for Neisseria
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

For confirmation of Campylobacter jejuni by means of the motility test.

Other Notes

Capability of selective media to detect heat-injured Shigella flexneri1

Price and Availability

Additional Products to Explore

for molecular biology

for microbiology

Sodium chloride

BioXtra, ≥99.5% (AT)

for microbiology

for use in microbial growth medium

Customers Also Viewed

for microbiology

for microbiology

for microbiology

for microbiology, ready to use media in the bottle

Microbiologically tested.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

On the trail of Campylobacter

The Campylobacter is one of the leading causes of human gastroenteritis. Common Campylobacter species C. jejuni, C. coli, and C. lari are responsible for most cases of campylobacteriosis1,2. However,...
Jvo Siegrist
AnalytiX Volume 8 Issue 1
Keywords: Antibiotics, Cancer, Citric Acid Cycle, Gram stain, Metabolism, Microbiology, Scanning electron microscopy

Streptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques

There are many natural sources of streptococci, including humans and diverse animals where they often colonize the mucosal surfaces of the mouth, intestinal tract, nasal passages and pharynx. The pre...
By Jvo Siegrist, Product Manager Microbiology, ivo.siegrist@sial.com
AnalytiX Volume 7 Article 3
Keywords: Diagnostic, Phosphorylations

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. J.L. Smith, B.J. Dell J. Food Prot. 53, 141, (1990)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?