26980 Fluka

Choline chloride

≥97.0% (AT)

DOWNLOAD MSDS (PDF)

Synonym: (2-Hydroxyethyl)trimethylammonium chloride

  • CAS Number 67-48-1

  • Linear Formula (CH3)3N(Cl)CH2CH2OH

  • Molecular Weight 139.62

  •  Beilstein Registry Number 3563126

  •  EC Number 200-655-4

  •  MDL number MFCD00011721

  • Popular Documents:  RAMAN FTIR (PDF)  

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
assay   ≥97.0% (AT)
mp   302-305 °C (dec.)(lit.)

Description

Substrates

Choline acetyltransferase substrate

Biochem/physiol Actions

The enzymatic activities of butyrylcholinesterase (BChE) and paraoxonase 1 (PON1), two serum enzymes synthesized by the liver and related with inflammation, were decreased in a sepsis animal model injected with LPS. Choline chloride administered intravenously at 20 mg/kg body weight prevents the LPS-mediated decreases in the activities of these two enzymes 1.

Price and Availability

Customers Also Viewed

Acetylcholine chloride

≥99% (TLC)

Choline chloride

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%

Choline chloride

≥99%

Choline chloride

≥98%

Choline chloride

BioUltra, ≥99.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?