27393 Fluka

1,4-Cineole

analytical standard, for terpene analysis

DOWNLOAD MSDS (PDF)

Synonym: 1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

  • CAS Number 470-67-7

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 104974

  •  EC Number 207-428-9

  •  MDL number MFCD00209502

  •  PubChem Substance ID 24856553

Properties

Related Categories A-F, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard, for terpene analysis
assay   ≥98.5% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.445(lit.)
  n20/D 1.445
bp   65 °C/16 mmHg(lit.)
mp   −46 °C(lit.)
density   0.887 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

1,4-Cineole

FG

1,4-Cineole

technical, mixture of isomers, ≥85% (GC)

1,8-Cineole
000205901,8-Cineole

primary reference standard

Eucalyptol

analytical standard, for terpene analysis

Eucalyptol

99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?