EMAIL THIS PAGE TO A FRIEND

29680 Fluka

Cyclopentane

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, C,
grade   analytical standard
vapor density   ~2 (vs air)
assay   ≥99.0% (GC)
form   neat
autoignition temp.   682 °F
shelf life   limited shelf life, expiry date on the label
expl. lim.   8.7 %
impurities   cyclopentene
refractive index   n20/D 1.405(lit.)
  n20/D 1.406
bp   50 °C(lit.)
mp   −94 °C(lit.)
density   0.751 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Cyclopentane

CHROMASOLV®, for HPLC, ≥75% cyclopentane basis

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
F
Risk Statements (Europe) 
Safety Statements (Europe) 
9-16-29-33-61
RIDADR 
UN 1146 3 / PGII
WGK Germany 
1
RTECS 
GY2390000
Flash Point(F) 
-2.2 °F
Flash Point(C) 
-19 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

QSPR modeling of octanol/water partition coefficient for vitamins by optimal descriptors calculated with SMILES. A A Toropov et al Eur. J. Med. Chem. 43, 714-40, (2008)

Loading...


Merck 14,2741

Beil. 5,IV,14

Aldrich MSDS 1, 530:D / Corp MSDS 1 (1), 984:B / FT-IR 1 (1), 33:C / FT-NMR 1 (1), 46:B / IR-Spectra (2), 24:G / IR-Spectra (3), 25:C / NMR-Reference 2 (1), 36:D / RegBook 1 (1), 33:B / Sax 6, 842 / Structure Index 1, 4:C:6 / Vapor Phase 3, 48:C

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?