Synonym:
4-
CAS Number 148476-30-6
Empirical Formula (Hill Notation) C17H20O3
Molecular Weight 272.34
MDL number MFCD10567354
| Related Categories | All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Chromatography, Environmental Standards, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?