30540 Fluka

Decane

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 124-18-5

  • Linear Formula CH3(CH2)8CH3

  • Molecular Weight 142.28

  •  Beilstein Registry Number 1696981

  •  EC Number 204-686-4

  •  MDL number MFCD00008954

  •  PubChem Substance ID 24858429

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, D,
grade   analytical standard
vapor density   4.9 (vs air)
vapor pressure   1 mmHg ( 16.5 °C)
  3.77 mmHg ( 37.7 °C)
assay   ≥99.8% (GC)
autoignition temp.   410 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   2.6 %
refractive index   n20/D 1.411(lit.)
  n20/D 1.412
bp   174 °C(lit.)
mp   −30 °C(lit.)
density   0.73 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Decane
457116Decane

anhydrous, ≥99%

Decane
30550Decane

purum, ≥98.0% (GC)

Decane
442669Decane

analytical standard

Decane
D901Decane

ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?