Synonym: Citric acid buffer solution
MDL number MFCD00146206
| Related Categories | Analytical Reagents, Analytical/Chromatography, Buffer Solutions, Buffer Solutions, pH 1 - 4, Titration, |
| quality | traceable to SRM from NIST and PTB |
| with fungicide | |
| pH | 1.99 (0 °C) |
| 1.99 (10 °C) | |
| 2.00 (20 °C) | |
| 2.00 (30 °C) | |
| 2.00 (40 °C) |
This material is tested against certified reference materials from NIST and/or PTB.
Visit our Titration Center to learn more.
Customers Also Viewed
hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB
di-sodium hydrogen phosphate / sodium hydroxide solution
with fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST
potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, traceable to SRM from NIST
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?