Synonym:
9-(2-Deoxy-β-
CAS Number 312693-72-4
Empirical Formula (Hill Notation) C10H13N5O4 · H2O
Molecular Weight 285.26
Beilstein Registry Number 39814
EC Number 213-505-8
MDL number MFCD00150760
| Related Categories | Biochemicals and Reagents, Nucleosides, Nucleosides, Nucleotides, Oligonucleotides More... |
| assay | ≥98.0% (HPLC) |
| solubility | 1 M NH4OH: soluble50 mg/mL, clear to very faintly turbid, yellow to brown |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?