Synonym: 2,2′,4,5,5′-PCB, 2,2′,4,5,5′-Pentachlorobiphenyl
CAS Number 37680-73-2
Empirical Formula (Hill Notation) C12H5Cl5
Molecular Weight 326.43
Ballschmiter Number 101
Beilstein Registry Number 2507418
EC Number 215-648-1
MDL number MFCD00055553
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Aroclors, PCBs, and Dioxins, Chromatography, |
| grade | analytical standard, for environmental analysis |
Customers Also Viewed
10 μg/mL each component in heptane, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?