Synonym:
2-Amino-4-tert-
CAS Number 30125-63-4
Empirical Formula (Hill Notation) C7H12ClN5
Molecular Weight 201.66
Beilstein Registry Number 514392
MDL number MFCD00144061
PubChem Substance ID 24862821
| Related Categories | All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Antibiotics, Chemical Structure, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?