31301 Fluka

Di-n-octyl phthalate

analytical standard, for environmental analysis

DOWNLOAD MSDS (PDF)

Synonym: DNOP, Phthalic acid di-n-octyl ester

  • CAS Number 117-84-0

  • Empirical Formula (Hill Notation) C24H38O4

  • Molecular Weight 390.56

  •  Beilstein Registry Number 1915994

  •  EC Number 204-214-7

  •  MDL number MFCD00015292

  •  PubChem Substance ID 24862966

Properties

Related Categories Allergens, Analytical Standards, Analytical/Chromatography, Chromatography, Cosmetic Standards,
grade   analytical standard, for environmental analysis
density   0.980 g/mL at 20 °C(lit.)

Price and Availability

Customers Also Viewed

Bis(2-ethylhexyl) phthalate

PESTANAL®, analytical standard

Di-n-octyl phthalate

≥98.0% (GC)

Diisodecyl phthalate

Selectophore, ≥99.0%

Diisononyl phthalate

ester content ≥99 % (mixture of C9 isomers), technical grade

Dioctyl phthalate

≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?