CAS Number 8049-11-4
Linear Formula AlCuZn
MDL number MFCD00198176
PubChem Substance ID 24858930
| Related Categories | Alloys, Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents, |
| grade | puriss. p.a. |
| form | powder |
| composition | aluminum, ~45% |
| copper, ~50% | |
| zinc, ~5% | |
| mp | 550 °C(lit.) |
Reducing agent used for the determination of total nitrogen by the method of Kjeldahl. Reagent used for the reduction of alkaline solutions of chlorate to chloride
reducing agent for the determination of nitrogen in alkaline pH-range
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?