Synonym: BDM, Biacetyl monoxime, Diacetyl monoxime
CAS Number 57-71-6
Linear Formula CH3C(=NOH)COCH3
Molecular Weight 101.10
Beilstein Registry Number 605582
EC Number 200-348-5
MDL number MFCD00002116
PubChem Substance ID 24858995
Popular Documents: FTNMR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Gravimetric Reagents, Spectroscopy, UV/Vis Reagents, |
| assay | ≥99.0% (N) |
| ≥99.0% | |
| quality | for spectrophotometric det. of urea |
| ign. residue | ≤0.05% (as SO4) |
| bp | 185-186 °C(lit.) |
| mp | 75-76 °C |
| 75-78 °C(lit.) |
Solubility:
0.5 g are completely soluble and give a clear solution in 10 mL water or also in 10 mL ethanol.
Sensitivity test:
0.05 mg urea in 3 mL water and 5 mL conc. HCl together with a 3% solution in water of diacetylmonoxime heated on a water bath for 10 minutes give a light yellow color.
Reagent for the colorimetric determination of urea and ureido-compounds1; Spectrometric reagent for Co(II), Ni(II), Pd(II) and Re(VII)2; Reagent for the gravimetric determination of Ni(II)3
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?