CAS Number 52918-63-5
Beilstein Registry Number 6746312
MDL number MFCD00870122
PubChem Substance ID 24869458
| grade | PESTANAL®, analytical standard |
| concentration | 100 ng/μL in cyclohexane |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?